Learn More
Fisherbrand™ Surface Tension Apparatus
Catalog No. 14818
Change view
Click to view available options
Includes:
250mm borosilicate glass capillary tube, 0 to 10cm, in 1mm divisions, cork holds capillary tube inside outer tube
Catalog No. Includes
14818 250mm borosilicate glass capillary tube, 0 to 10cm, in 1mm divisions, cork holds capillary tube inside outer tube
1 options
Catalog No. 14818 Supplier Fisherbrand™ Supplier No. 14818
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Determines surface tension from height of liquid in capillary tube.
  • 250mm borosilicate glass capillary tube
  • Engraved graduations
  • Graduated from 0 to 10cm, in 1mm divisions
  • Cork holds capillary tube inside outer tube
  • Outer tube accommodates rubber tubing
Specifications
Product Type Surface Tension Apparatus
Includes 250mm borosilicate glass capillary tube, 0 to 10cm, in 1mm divisions, cork holds capillary tube inside outer tube
For Use With (Application) Determines surface tension from height of liquid in capillary tube