Learn More
Fisherbrand™ SureStain™ Stains
Description
- For manual staining
- Produce uniform results
- Use for immersion or rack-type staining procedures
- Entire procedure can be performed in 30 seconds
Order Info
32 oz. bottles.
Specifications
Specifications
| Quantity | 32 oz. |
| Type | Wright-Giemsa Stain |
| For Use With (Application) | For differential blood cell analysis |