Learn More
Fisherbrand™ SureStain™ Stains
Fast, reliable staining for differential blood cell analysis
Supplier:  Fisherbrand™ CS432
 View more versions of this product
Catalog No. CS432D
Only null left
Description
Description
  • For manual staining
  • Produce uniform results
  • Use for immersion or rack-type staining procedures
  • Entire procedure can be performed in 30 seconds
Order Info
32 oz. bottles.
Specifications
Chemical Identifiers
32 oz.
Specifications
32 oz.
Wright Stain
Fast, reliable staining for differential blood cell analysis
Product Suggestions
Product Suggestions