Learn More
Fisherbrand™ Stainless-Steel Racks for Quartz Crucibles for Isotemp Furnaces
Rack keeps crucibles firmly in place while in the Isotemp furnace.
Supplier:  Fisherbrand™ 52682
 View more versions of this product
Catalog No. 104975
Only null left
Description
Description
For use with 10-490-4 (25mm diameter) crucibles in all Isotemp furnaces.
  • Handle for racks (10-490-17) sold separately; order online
Specifications
Specifications
Rack for 15 crucibles
Product Suggestions
Product Suggestions