Learn More
Fisherbrand™ Sound Enclosure for Model 50 and 120 Sonic Dismembrator
Catalog No. FB432A
Change view
Click to view available options
For Use With (Equipment):
Model 50/120 Sonic Dismembrator
Catalog No. For Use With (Equipment)
FB432A Model 50/120 Sonic Dismembrator
1 options
Catalog No. FB432A Supplier Fisherbrand™ Supplier No. FB432A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sound Enclosure for Model 50/120 Sonic Dismembrator. Sonic Dismembrators are extremely loud devices that can cause discomfort to the user and anyone nearby. The Sound Enclosure reduces noise by approximately 20 dBa and is made to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Fisherbrand probe or horn will be held safely and securely inside the unit.
This Fisher Scientific™ Sound Enclosure is designed for use with Model 50 and Model 120 Sonic Dismembrators
  • Sound Reducing Enclosure including stand and clamp
  • For use with Model 50/120 Sonic Dismembrator
  • Two ports are located on either side of the enclosure for coolant tubing or a temperature monitoring probe
  • Interior walls are lined with water-resistant acoustical foam
  • Door has a window so experiments can be visually monitored
  • Dimension: 50.8 x 25.4 x 25.4cm (20 x 10 x 10 in.)
Specifications
Product Type Sound Enclosure
Dimensions (L x W x H) 12 x 12 x 20 in.
Height (English) 20 in.
For Use With (Application) Ideal for sound reduction
For Use With (Equipment) Model 50/120 Sonic Dismembrator
Includes Internal support rod and converter mounting system