Learn More
Fisherbrand™ Set of Eight Precision Specific Gravity Hydrometers
Specific Gravity Hydrometers are permanently marked with individual serial numbers.
$5,082.61
Specifications
| Length (Metric) | 300 mm |
|---|---|
| For Use With (Equipment) | 300mL Borosilicate Glass cylinder |
| Material | Soda Lime Glass |
| Temperature Range (English) | 0°F to 220°F |
| Specific Gravity Range | 0.695 to 2 |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste.
Description
Specific Gravity Hydrometers are permanently marked with individual serial numbers.
Set includes thermometer with 0° to 220°F range and eight precision hydrometers that cover a specific gravity range of 0.695 to 2.000.
- Approximately 300mm long for use with 08-530J cylinder, available separately
- In lined case
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST™.
Compliance
Accurate to one graduation line that meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM™ E 126 specifications.
Specifications
| 300 mm | |
| Soda Lime Glass | |
| 0.695 to 2 |
| 300mL Borosilicate Glass cylinder | |
| 0°F to 220°F |