Learn More
Fisherbrand™ Power Hold Universal Clamp for Overhead Stirrers
Can be used for most applications.
$181.41
Specifications
Size Universal
Type Clamp
Material Cast Zinc, Aluminum
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
14512906
View Documents Promotion Details
Fisherbrand™
370209046
Stirring stand fr. 9.5 to 16 mm
Each for $181.41
Only null left
14512907
View Documents Promotion Details
Fisherbrand™
370209047
Stirring stand fr. 12 to 19 mm
Each for $181.41
Only null left
Description
Description
  • Designed with large knobs for powerful gripping
  • Adjustable stop for speed and safety in set up and operation
  • Made with precision machined aluminum rod for strength and durability
Specifications
Specifications
Universal
Clamp
Cast Zinc, Aluminum