missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MultiTube Vortexers: Replacement Tray Pad Set

Catalog No. 02215460
Change view
Click to view available options
For Use With (Equipment):
Multitube vortexer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
02215460 Multitube vortexer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02215460 Supplier Fisherbrand™ Supplier No. 80862907

Replacement parts for Fisherbrand™ MultiTube Vortexers.

For use with Fisher Scientific MultiTube Vortexers

  • Replacement tray pad set includes top and lower pads
  • Gray

Specifications

Product Type Tray Pad Set
Color Gray
For Use With (Equipment) Multitube vortexer