missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mini-Tube Rotator Accessories
GREENER_CHOICE

Interchangeable rotisserie options for all common laboratory tubes (1.5/2.0, 5/7, 15 and 50mL tubes)

Supplier:  Fisherbrand™ 260752

 View more versions of this product

Catalog No. 05450129


Only null left

Description

Description

Fisherbrand mini-tube rotator accessory carousels are available in tube capacities 36 of 1.5 to 2mL, 10 of 10/15mL and 16 of 5/7mL, and 6 of 50mL.

  •  Ideal for mixing laboratory tubes
  • Cold room and incubator compatible
  • Adjustable 0 to 90 degrees
  • Durable material
  • 10 of 10/15mL and 16 of 5/7mL tubes
Specifications

Chemical Identifiers

Carousel
 Mixing
 1EA

Specifications

5 to 15 mL
 Mixing
10
Carousel
 1EA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions