Learn More
Fisherbrand™ Midget Urinometer
Catalog No. 22274209
Change view
Click to view available options
Accuracy:
±0.002
Catalog No. Accuracy
22274209 ±0.002
1 options
Catalog No. 22274209
Urinometer accuracy guaranteed to ±0.002.
  • Set consists of a glass jar and standard shot ballast hydrometer with smooth outside wall
  • Range of measurements is from 1.000 to 1.040 in 0.001 increments
  • Replacement hydrometer and jar available separately
Specifications
Accuracy ±0.002
Increments 0.001
Length (Metric) 110 mm
Includes Glass jar and standard shot ballast hydrometer with a smooth outside wall
Material Soda Lime Glass
For Use With (Application) Urine analysis