missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxima™ C Plus Pump Oil

Catalog No. p-159713
Change view
Click to view available options
Volume (Metric):
0.95 L
4 L
19 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
012574D 19 L
012574B 4 L
012574A 0.95 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 012574D Supplier Fisherbrand™ Supplier No. 012574D
Only null left

Triple-distilled hydrocarbon pump oil designed to resist breakdown in Fisher Scientific Maxima C Plus pumps.

Designed to resist breakdown in Fisher Scientific Maxima C Plus pumps

Triple-distilled hydrocarbon oil using severely hydrotreated base stock. Hydrotreating minimizes aromatics and sulfur to give good resistance to sludge and varnish formation over time in corrosive environments. The oil enables pumps to maintain the highest vacuum performance over time. Clear, colorless, and odorless.

Specifications

Product Type Pump Oil
For Use With (Application) Oil enables pumps to maintain the highest vacuum performance over time
For Use With (Equipment) Maxima™ C plus vacuum pump series
Color Clear
Quantity 1 Ea.
Volume (English) 5.01 gal.
Volume (Metric) 19 L