Learn More
Fisherbrand™ Jack Stand
Catalog No. FB357
Change view
Click to view available options
For Use With (Equipment):
Model 505/705 Sonic Dismembrator
Catalog No. For Use With (Equipment)
FB357 Model 505/705 Sonic Dismembrator
1 options
Catalog No. FB357 Supplier Fisherbrand™ Supplier No. FB357
Ideal for raising samples up to a stationary probe mounted within the enclosure or on a stand.
Fisher Scientific™ Jack Stand is ideal for samples to a stationary probe mounted within the enclosure.
  • Platform that raises and lowers samples under a stationary probe
  • For use with Model 505/705 Sonic Dismembrator.
Specifications
Product Type Jack Stand
Dimensions (L x W) 6 x 6 in. platform
For Use With (Application) Ideal to raise samples to a stationary probe mounted in an enclosure
For Use With (Equipment) Model 505/705 Sonic Dismembrator
Packaging Quantity 1 Ea.