Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Supplier: Fisherbrand™ 80863642
Description
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Accessories Offered:
- Well plate blocks with lid
- PCR plate blocks with lid
- Tube blocks
- Conical tube blocks
- Microplate thermal blocks
Chemical Identifiers
| Shake Touch Shakers |
Specifications
| 384-well Plate | |
| 0.32 in. | |
| 0.16 in. | |
| 4.7 x 6.4 x 3.0 in. (11.9 x 16.3 x 7.6 cm) | |
| Shake Touch Shakers |
| 384-well Plate Block with Lid | |
| 0.81 cm | |
| 0.4 cm | |
| Heating/Shaking | |
| 384 |