Learn More
Fisherbrand™ Isotemp™ Refrigerator and Freezer Alarm Systems
Catalog No. 13986100
Change view
Click to view available options
Deluxe remote system
Catalog No.
13986100 Deluxe remote system
1 options
Catalog No. 13986100 Supplier Fisherbrand™ Supplier No. 13 986 100
Requires
Remote systems require an existing alarm system, with dry alarm contacts, to be installed on the refrigerator or freezer in order to be compatible.
For Isotemp and Isotemp Plus™ refrigerators and freezers
Freestanding Alarm System
  • Use with basic freezers, refrigerators, and one section on refrigerator/freezer combinations With key switch in ALARM position, system provides audio and visual alarms if chamber temperature reaches high or low alarm limit (adjustable, set by user) or if power is lost for more than 30 seconds
  • Housed in a painted metal cabinet with digital temperature display
  • Alarm silencing and test features
  • Can be used to trigger Standard or Deluxe Remote Alarm systems
Standard Remote System†
  • Use with refrigerators and freezers equipped with panel-mounted or freestanding alarm (Alarm or Recorder/Alarm models)
  • Can be located up to one-third mile from unit
  • Emits audible and visual signals in the event of power failure, temperature deviation, or if door is open for more than three minutes
  • Alarm sounds if line between unit and alarm is cut
Deluxe Remote System†
  • Use with refrigerators and freezers equipped with panel-mounted or freestanding alarm (Alarm or Recorder/Alarm models)
  • Can monitor up to four individual or four groups of refrigerators or freezers
  • User-programmable to call up to eight telephone or pager numbers when activated by power failure, temperature deviation, or if door is open for more than three minutes
  • Alarm sounds if line between unit and alarm is cut
  • Stores four outgoing messages
  • Automatic redial for busy or unanswered numbers
  • Works with any standard telephone system
  • Battery backup allows alarm to sound in the event of a power failure
Order Info
Freestanding and remote systems must be ordered when the refrigerator or freezer is ordered. Installation by owner.
† Requires an existing alarm system, with dry alarm contacts, to be installed on the refrigerator or freezer in order to be compatible.
Specifications
Type Deluxe Remote System
For Use With (Equipment) Isotemp™ and Isotemp™ Plus™ refrigerator and freezer