missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Large-Capacity Refrigerated Incubator Chart Recorder Paper

Catalog No. 1507480
Change view
Click to view available options
Quantity:
60 Sheets/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1507480 60 Sheets/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1507480 Supplier Fisherbrand™ Supplier No. C653

For Use With: Large-Capacity Refrigerated Incubator in temperates between 0°C to +50°C

  • Circular, seven-day, 6 in. diameter (15cm)
  • Range is user determinable
  • Comes in package of 60 sheets

Specifications

For Use With (Equipment) Large-Capacity Refrigerated Incubator
Temperature Range (Metric) 0°C to 50°C
Type Chart Recorder Paper
Quantity 60 Sheets/Pk.
Diameter (English) Base 6 in.
Diameter (Metric) Base 15 cm