missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Concentric Ring Cover for Precision™ Water Baths, 6 Holes, for 20L and 28L Water Bath

These Fisherbrand™ Covers are for use with Precision Dubnoff and Shallow-Form Shaking Water Baths.

Supplier:  Fisherbrand™ 1546231Q

Catalog No. 1546231Q

Only null left

Description

Description

These Fisherbrand™ Covers are for use with Precision Dubnoff and Shallow-Form Shaking Water Baths.

Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods

Warranty

1 Year

Specifications

Specifications

Concentric Ring Cover 6 Holes
Precision 20L and 28L General Purpose Water Baths
6
20, 28 L
Stainless Steel
1 Year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions