missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters

Catalog No. 88860023
Change view
Click to view available options
No. of Places:
1
2
4
Plug Type:
US
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Places Plug Type
88860023 4 US
88860021 1 US
88860022 2 US
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 88860023 Supplier Fisherbrand™ Supplier No. 88860023

Requires

Unit must be ordered with block(s)

Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Warranty

2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID
Material Powder-coated Steel
No. of Places 4
Plug Type US
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C
Type Dry Bath Block
Fuse 250V 2.5A
Heat Up Time ≤ 20 min. 30° to 130°C
Includes Dry bath, US style plug, block lifter
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 1°C
Timer 0 to 99:59 min or continuous
Warranty 2 Year, Replacement
Dimensions (L x W x H) 17.7 x 7.9 x 3.9 in. (450 x 200 x 100 mm)
Length (English) 17.7 in.
Length (Metric) 450 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 3.9 in.
Height (Metric) 100 mm
Weight (English) 10.36 lb.
Weight (Metric) 4.7 kg
Electrical Requirements 100/120 V, 50/60 Hz
Voltage 100/120 V
Frequency 50/60 Hz
Show More Show Less