Learn More
Fisherbrand™ Electrode Holder
Catalog No. 22286439
Catalog No. 22286439
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Handles multiple pH and conductivity readings quickly with reduced electrode breakage. Fisherbrand Electrode Holder accepts all brands of standard electrodes; extends 21 in. and pivots 360°; designed for smooth movement and effortless operation.
  • Articulated at three points, the arm holder pivots in any direction (360°) while holder maintains electrode in vertical position
  • Extends to 21 in. (53cm) from weighted 8 in. (20cm), die-cast metal base
  • Accommodates standard glass, reference, and combination electrodes as well as temperature and conductivity probes
  • Spring counterbalance for smooth, fingertip control and stability
Specifications
Type Holder
For Use With (Application) pH