missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boekel Scientific™ Dricycler
Description
- Automatically regenerates desiccant every five hours
- Adjustable shelves
- Supplied with 2 shelves
- Hand crafted from stainless steel and glass
Specifications
Specifications
| Cabinet Style | Front-loading |
| Material | Stainless Steel, Glass |
| Product Type | Dricycler |
| For Use With (Application) | Storing reagents or soil samples, drying biological specimens, storage sludge samples, cooling down from 40° to 150°C. |
| Includes | Adjustable shelves, neoprene door gasket, full-length door hinges |
| No. of Shelves | 2 |
| Shelf Positions | 1/2 in. Increments |
| Size | Small |
| Warranty | 2 Years |
| Dimensions (L x W x H) | 12.5 x 16 x 12.25 in. (31.4 x 40.6 x 31 cm) |
| Show More |