missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Digital Key-Chain Wristband Counter

Lightweight and easy to use Digital Key-Chain Wristband Counter.

Supplier:  Fisherbrand™ 3129EMD

Catalog No. S94108

Only null left
Explore available promotions

Description

Description

Easy-to-read LCD counts to 99,999 and comes with a two-year constant-use battery. Reduces miscounting problems associated with mechanical counting equipment.
  • ABS plastic, O-ring sealed case is water and shock resistant
  • Equipped with button click for count confirmation
  • Offers recessed 'no accident' reset
  • Diameter x H: 1.88 x 0.5 in. (47 x 13 mm)
Specifications

Specifications

Digital Counter
99,999
1 Ea.
4.7 cm
1.27 cm
2 Years constant use battery
LCD
ABS Plastic
1.8 in.
0.5 in.
Ease-of-use and fail-safe counting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions