Learn More
Fisherbrand™ Baume Hydrometers
Baume heavy scale for working with liquids heavier than water.
Description
Baume heavy scale for working with liquids heavier than water.
- Permanently marked with individual serial numbers
Order Info
Use Fisherbrand cylinder 08-530J with 250mL of liquid for submersion, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
Chemical Identifiers
| Baume Hydrometer |
Specifications
| Baume Hydrometer | |
| Without 3-point certification | |
| Soda Lime Glass | |
| Baume 'Heavy' Scale For Liquids Heavier than Water |
| 0.1° | |
| 290 mm | |
| 40 to 51° |