Learn More
Fisherbrand™ Baume Hydrometers
Baume heavy scale for working with liquids heavier than water.
 View more versions of this product
Catalog No. 11571A
Only null left
Description
Description
Baume heavy scale for working with liquids heavier than water.
  • Permanently marked with individual serial numbers
Order Info
Use Fisherbrand cylinder 08-530J with 250mL of liquid for submersion, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
Specifications
Chemical Identifiers
Baume Hydrometer
Specifications
Baume Hydrometer
Without 3-point certification
Soda Lime Glass
Baume 'Heavy' Scale For Liquids Heavier than Water
0.1°
310 mm
0 to 12°
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories