Learn More
Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
$940.35
Specifications
Display Analog
Voltage 0/1 V
For Use With (Application) Safely mixing all types of solvents, lacquers and volatile chemicals
Includes 10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
Material Steel Aluminum
View More Specs
Includes:
  • A 9/16 in. dia. x 10 in. L support rod
  • Four ft. air hose with double fittings for connection to air supply
Products 1
Catalog Number Mfr. No. Speed Capacity Torque Price Quantity & Availability  
Catalog Number Mfr. No. Speed Capacity Torque Price Quantity & Availability  
14500219
View Documents Promotion Details
Fisherbrand™
370209017
10,000 rpm 10 L 45
Each for $940.35
Only null left
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Order Info
Shaft, propeller, stand and clamp available separately
Specifications
Specifications
Analog
Safely mixing all types of solvents, lacquers and volatile chemicals
Steel Aluminum
0/1 V
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
Air Powered Overhead Stirrer