missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Submersible pH Combination Electrode

Catalog No. 13620AP56
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13620AP56 BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13620AP56 Supplier Fisherbrand™ Supplier No. 3580523
Only null left

Submersible pH electrode features large backlit LCD showing the measured value, measured units, temperature, time, battery life and menu options.

  • Large backlit LCD shows the measured value, measured units, temperature, time, battery life and menu options
  • pH calibration capabilities include up to six calibration points
  • User-settable calibration alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN and PWB standards
  • Set point alarms indicate when concentrations are outside of programmed limits
  • Salinity correction is automatically compensated for after manual input of concentration

Warranty

One year

Specifications

Connector Types BNC
Junction Single
Material ABS
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 80°C
Type pH submersible Combination Electrode
Filling Solution Type Gel
For Use With (Application) Field
For Use With (Equipment) pH meter
pH Relative Accuracy ±0.01
Temperature Accuracy ±0.3°C
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Length (English) Cable 120 in.
Length (Metric) Cable 3 m
Diameter (English) 1 in.
Diameter (Metric) 25 mm
Show More Show Less