Learn More
Fisherbrand™ accumet™ Replacement Conductivity/TDS/Temperature Probe - Mercury - Free
Replacement probe to be used for conductivity, TDS, and temp.
Supplier:  Fisherbrand™ 3560850DEL
Catalog No. 13-620-AP54
Only null left
Description
Description
Replacement probe to be used for conductivity, TDS, and temp.
Replacement for:
  • accumet AP75, AP85 Portable Meters
  • Oakton™ CON 11, CON 110, CON 400, CON 410, pH/CON 300, TDS 400 Portable Meters
  • Oakton CON 510, pH/CON 510 Benchtop Meters
Specifications
Specifications
Conductivity, TDS, Temp
Accumet AP75, AP85 portable meter, Oakton CON 11, CON 110, CON 400, CON 410, pH/CON 300, TDS 400 portable meter, Oakton CON 510, pH/CON 510 benchtop meter
0.9 m
Conductivity/TDS/Temperature
Conductivity/TDS/Temperature Probe
36 in.
Epoxy
Product Suggestions
Product Suggestions