missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ pH/ATC Probe for Portable AP80 and AP70 Series Meters: Mercury-Free

Catalog No. 13620AP55
Change view
Click to view available options
Connector Types:
BNC, ATC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13620AP55 BNC, ATC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13620AP55 Supplier Fisherbrand™ Supplier No. 13 620 AP55
Only null left

Replacement/accessory for accumet AP85, AP84, AP72, AP71 Portable Meters

  • All-in-one pH/temperature probe
  • Liquid-filled Ag/AgCl reference element
  • Rugged plastic body

Specifications

Connector Types BNC, ATC
Junction Single
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +80°C
Type pH/ORP Combination Titration Electrode
Filling Solution Type Saturated KCl with AgCl. KCl
For Use With (Application) All-in-one pH temperature
For Use With (Equipment) Accumet AP70 portable meter
Internal Reference Ag/AgCl
Temperature (English) Operating 23°F to 176°F
Warranty 1 Year
Show More Show Less