missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

$891.32

Specifications

Junction Single
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +110°C
Filling Solution Type Saturated KCl with AgCl. KCl
View More Specs

Products 2
Catalog Number Mfr. No. Connector Types Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Length (Metric) Price Quantity & Availability  
13620291
Accessories View Documents Promotion Details
Fisherbrand™
13 620 291
BNC 100 mm
Each for $891.32
Only null left
 
13620292
Accessories View Documents Promotion Details
Fisherbrand™
13 620 292
BNC 150 mm
Each for $891.32
Only null left
 
Description

Description

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

Zero to 14pH range
Specifications

Specifications

Single
0 to 14
Saturated KCl with AgCl. KCl
Ag/AgCl
-5°C to +110°C
1 Year
Glass
-5°C to +110°C
Small samples
23°F to 230°F
Micro
Videos
SDS
Documents

Documents