missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Micro Glass Mercury-Free Combination Electrodes

Catalog No. 13620851
Change view
Click to view available options
Length (English):
5 in.
10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
13620851 10 in.
13620850 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13620851 Supplier Fisherbrand™ Supplier No. 13620851

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes

  • Glass body (3 x 38mm) with 3mm teflon stem
  • Refillable
  • TRIS compatible
  • Mercury-free
  • Double junction
  • pH Range: 0 to 14

Specifications

Connector Types BNC
Junction Double
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
For Use With (Application) Capillary Tubes, Cuvettes, Microwell Plates, Micro Test Tubes, NMR Tubes, Serum Cups
Temperature (English) Operating 32°F to 176°F
Length (English) 10 in.
Length (Metric) 25.4 cm
Length (Metric) Stem 15.2 cm
Diameter (English) 0.12 in.
Diameter (Metric) 3 mm
Show More Show Less