missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Micro Glass Mercury-Free Combination Electrodes
Description
- Glass body (3 x 38mm) with 3mm teflon stem
- Refillable
- TRIS compatible
- Mercury-free
- Double junction
- pH Range: 0 to 14
Specifications
Specifications
| Connector Types | BNC |
| Junction | Double |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | Combination Electrode |
| For Use With (Application) | Capillary Tubes, Cuvettes, Microwell Plates, Micro Test Tubes, NMR Tubes, Serum Cups |
| Temperature (English) Operating | 32°F to 176°F |
| Length (English) | 5 in. |
| Length (Metric) | 12.7 cm |
| Show More |