missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Liquid-Filled Mercury-Free pH/ATC Epoxy Body Combination Electrodes, w/BNC and Mini-phone ATC connectors

Catalog No. 13620AP61
Change view
Click to view available options
Temperature (Metric) Operating:
0°C to 100°C
0°C to 80°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature (Metric) Operating
13620AP61 0°C to 100°C
13620631 0°C to 80°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13620AP61 Supplier Fisherbrand™ Supplier No. 13 620 AP61
Only null left

Includes

Electrode, 60mL refill solution, electrode storage bottle

Requires

Requires filling solution

TRIS compatible, rugged, and convenient

  • Rugged standard-size epoxy body
  • Double-junction design prevents reference contamination; compatible with TRIS applications; mercury free
  • Liquid filled design provides quick response
  • Porous ceramic junction, Ag/AgCl reference
  • Built-in ATC probe

  • Rugged epoxy body for use in field or plant
  • Ideal for handheld use with aqueous samples
  • Double-junction design eliminates measurement problems and clogging in research with TRIS buffers

Order Info

Endcap seal; keeps membrane moist and ready

Specifications

Color Purple
Connector Types BNC/Mini-Phone WP-ATC
Junction Porous Ceramic Double
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type pH/ATC Combination Titration Electrode
Filling Solution Type Saturated KCl
For Use With (Application) General laboratory and life science applications
For Use With (Equipment) Accumet AP60 and AP100 portable meter
Includes Electrode, 60mL refill solution, electrode storage bottle
Internal Reference Ag/AgCl
Probe ATC
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.1 in.
Length (Metric) 105 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less