missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, 0.100 M (M/10), Fisherbrand
Ammonium chloride
Supplier: Ricca Chemical Company ACCU0800500A
Description
Ammonium chloride. 500ml total volume sample with a concentration of 0.1M
- 0.1 M Concentration
- Ammonia ISE
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
0.1 M | |
Standard | |
Natural Poly Bottle | |
500 mL |
Ion Selective Electrode | |
Ammonium Chloride | |
500 mL |