Learn More
Fisherbrand™ accumet™ Automatic Temperature Compensation Probes - Mercury - Free
Description
For use with accumet Meters
- Signal solution temperature to meter for automatic compensation
- Permit temperature readout with meters having this feature
- Probes for accumet AP Series portable meters have waterproof connector
Specifications
Specifications
| Material | Stainless Steel |
| Type | ATC |
| For Use With (Equipment) | Accumet model AP63, AP62, AP61 |
| Connector Types | Mini-Phone |
| Length (Metric) Cable | 0.9 m |