missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AP115 Portable pH Meter Kit

Catalog No. 13636AP115
Change view
Click to view available options
Range (pH):
-2 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13636AP115 -2 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13636AP115 Supplier Fisherbrand™ Supplier No. 13636AP115
  • ONLINE EXCLUSIVE PRICE
    $2,467.63
    $1,186.32 / Each

    Save $1,281.31

    52.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Meter, accumet combination refillable pH/ATC electrode 13-620-AP50A, refill solution, hard carrying case, sample bottles, pH calibration packets, 9V battery and manual.

Accurate measurements any time, anywhere

For indoor and outside use. Versatile and rugged enough for the most demanding applications. Soft touch keys with comfort grip make one-handed operation a breeze. Compact design is small enough for a shirt pocket yet smart enough for any laboratory. Kit includes an unbreakable accumet pH/ATC electrode that is refillable for fast response and longest life.

AP115 pH/mV/Temp meter offers all features of the AP110 meter, plus:

  • 0.001pH resolution
  • Date/time display to meet GLP requirements

Specifications

Includes Meter, Accumet™ combination refillable pH/ATC electrode 13-620-AP50A, refill solution, hard carrying case, sample bottles, pH calibration packets, 9V battery and manual
For Use With (Application) pH, mV, Temperature
Measurement Modes pH, Relative mV, Temperature
Calibration pH, Relative mV, Temperature
Range (pH) -2 to 20
Resolution (pH) 0.001
Range (mV) 0 to ±2000
Calibration Points 1 to 5 pH, 1 RmV, 1 temperature
Temperature Range (Metric) -5°C to +100°C
Display Type LCD
Backlight Option Yes
Data Points 200
Log Function Type Automatic, Manual
Certifications/Compliance GLP
Warranty 3 Years
Accuracy Multiple
Resolution 0.001pH
Detection Range -2 to 20 pH
Type AP115 Portable pH Meter Kit
Show More Show Less