Learn More
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator
Catalog No. FB4103
Change view
Click to view available options
Product Type:
Temperature Probe
Thermocouple Probe
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
FB4103 Thermocouple Probe
FB4102 Temperature Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FB4103 Supplier Fisherbrand™ Supplier No. 4103
For monitoring temperature of sensitive samples
  • For monitoring temperature sensitive samples volumes
  • Can be used to monitor volumes from 1 - 1000mL
  • Temperature monitoring thermocouple bead on the end of a hermetically sealed wire
Specifications
Product Type Thermocouple Probe
For Use With (Application) Ideal for monitoring temperature of samples >1mL and indirect sonication methods
For Use With (Equipment) FB 705 Sonic Dismembrator™