missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Pencils

Catalog No. S04191
Change view
Click to view available options
Color:
Black
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
S04191 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04191 Supplier Fisher Science Education™ Supplier No. PL5000W

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commonly used in the science classroom.

Specifications

Product Type Pencil
Color Black
For Use With (Application) Used in the science classroom
Quantity Each