missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Foam Tray

Catalog No. S04188
Change view
Click to view available options
Dimensions (L x W):
9 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S04188 9 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04188 Supplier Fisher Science Education™ Supplier No. ST5003P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Multi-use tray commonly used in the science classroom.

  • Multi-use tray for classrooms
  • 9.25 x 12.13 x 0.75
  • Made of sturdy foam material
  • Disposable for easy clean-up

Specifications

Product Type Tray
Color White
Material Foam
For Use With (Application) For science classroom
Dimensions (L x W) 9 x 12 in.