Learn More
Fisher Science Education™ Elementary Zoom Microscope
Catalog No. S19210
Change view
Click to view available options
Magnification Power:
25X to 100X
Catalog No. Magnification Power
S19210 25X to 100X
1 options
Catalog No. S19210 Supplier Fisher Science Education™ Supplier No. 2056 EMD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Easy zoom focuses 25 to 100X!
An excellent starter microscope for elementary students, or field work where electricity is not available. Easily focus and zoom from 25X to 100X by rotating the viewing tube. Light is provided by the reversible mirror, concave on one side, and white diffusion space on the other side. Features achromatic polished glass lenses.
Specifications
Eyepiece WF 10X
Lighting Type Mirror
Includes Tamper proof viewing tube, shock resistant stand, achromatic ground and polished Glass lenses, permanently attached reversible mirror, dust cover and instruction manual
Magnification Power 25X to 100X
For Use With (Application) Elementary students or field work where electricity is not available