missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ Sure-Vue™ Adeno Test Kit

Catalog No. SA067020
Change view
Click to view available options
Quantity:
20 Tests
Type:
Adenovirus Test Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
SA067020 20 Tests Adenovirus Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA067020 Supplier Fisher Healthcare™ Supplier No. 067020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

20 Test devices, tubes containing extraction buffer, disposable sample transfer pipets, package insert

Simple two-step procedure

  • CLIA Complexity: Moderate
  • CPT Code: 87301
  • Results in 15 minutes
  • Rapid, membrane-based immunogold assay for the qualitative detection of adenovirus and its antigens
  • Aids in the diagnosis of conjunctivitis, respiratory illnesses and gastroenteritis caused by adenovirus
  • Can be used with eye swabs, nasopharyngeal secretions, fecal samples, and cell culture supernatant
  • Positive and negative controls sold separately

Specifications

CPT Code 87301
Sample Type Cell Culture Supernatant, Eye Swab, Fecal, Nasopharyngeal Secretions
Type Adenovirus Test Kit
For Use With (Application) Qualitative detection of Adenovirus and its Antigens in eye swabs, nasal and pharyngeal secretions and fecal material
Includes 20 Test devices, tubes containing extraction Buffer, disposable sample transfer pipets, package insert
Quantity 20 Tests
Test Time 15 min.