missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Stains for HEMA-TEK™ Automated Slide Stainers

Catalog No. 23036221
Change view
Click to view available options
Quantity:
5 Case
Each
Color:
Dark Blue
Dark Purple
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
23036221 Each Dark Blue
23036222 Each Dark Purple
23036507 5 Case Dark Blue
23036506 5 Case Dark Purple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23036221 Supplier Fisher Healthcare™ Supplier No. 036221
Only null left

Allow varied staining times to suit individual preferences

Stains for manual and automated staining methods

  • All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
  • Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
  • Carefully balanced for optimum staining pH
  • Remain stable for a minimum of 12 months when stored at room temperature
  • Packaged in durable plastic bottles for safety and easy handling
  • Convenient finger tabs and clear reagent identification eliminate improper line connection

Specifications

Quantity Each
Description Stain, Wright-Giemsa Stain
Storage Requirements Room Temperature
Boiling Point 65°C
Format Liquid
For Use With (Equipment) Hema-Tek™ automated slide stainer
Color Dark Blue