missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains

Catalog No. 23314631
Change view
Click to view available options
Quantity:
475 mL
500 mL
1 L
1 gal.
4 L
Product Type:
Alcoholic Eosin Y Stain
Bluing Reagent
Cyto-Quik Combo EA/OG Stain
Eosin Y Intensified
Gill Hematoxylin (1X)
Gill Hematoxylin (2X)
Gill Hematoxylin (3X)
Gomori Trichrome
Harris Hematoxylin
Harris Hematoxylin - Mercury-free
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23314631 1 gal. Eosin Y Intensified
23245651 4 L Harris Hematoxylin - Mercury-free
23245655 4 L Gill Hematoxylin (2X)
23245653 4 L Gill Hematoxylin (1X)
23245658 1 L Alcoholic Eosin Y Stain
23245657 4 L Gill Hematoxylin (3X)
23245654 475 mL Gill Hematoxylin (2X)
23245681 4 L Bluing Reagent
23245678 4 L Harris Hematoxylin
23245679 500 mL Gomori Trichrome
23245827 4 L Alcoholic Eosin Y Stain
23310349 1 gal. Cyto-Quik Combo EA/OG Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 23314631 Supplier Fisher Healthcare™ Supplier No. 314631

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured with strict quality control to ensure lot-to-lot uniformity

Prepared with stain certified by the Biological Stain Commission

Specifications

Format Liquid
For Use With (Application) Used for Histological/Cytological Staining
Quantity 1 gal.
Product Type Eosin Y Intensified