missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains

Catalog No. 23245827
Change view
Click to view available options
Quantity:
475 mL
500 mL
32 oz.
1 L
1 gal.
4 L
Type:
Alcoholic Eosin Y Stain
Bluing Reagent
Cyto-Quik Combo EA/OG Stain
Eosin Y Intensified
Gill Hematoxylin (1X)
Gill Hematoxylin (2X)
Gill Hematoxylin (3X)
Gomori Trichrome
Harris Hematoxylin
Harris Hematoxylin - Mercury-free
Harris Hematoxylin - Mercury-free, Non-acidified
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23245827 4 L Alcoholic Eosin Y Stain
23245651 4 L Harris Hematoxylin - Mercury-free
23245655 4 L Gill Hematoxylin (2X)
23245653 4 L Gill Hematoxylin (1X)
23245658 1 L Alcoholic Eosin Y Stain
23245657 4 L Gill Hematoxylin (3X)
23245654 475 mL Gill Hematoxylin (2X)
23245681 4 L Bluing Reagent
23245678 4 L Harris Hematoxylin
23245677 475 mL Harris Hematoxylin - Mercury-free
23245679 500 mL Gomori Trichrome
23245697 4 L Harris Hematoxylin - Mercury-free, Non-acidified
23310349 1 gal. Cyto-Quik Combo EA/OG Stain
23314631 1 gal. Eosin Y Intensified
23314630 32 oz. Eosin Y Intensified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 23245827 Supplier Fisher Healthcare™ Supplier No. 245827
Only null left

Manufactured with strict quality control to ensure lot-to-lot uniformity

Prepared with stain certified by the Biological Stain Commission

Specifications

Type Alcoholic Eosin Y Stain
Format Liquid
For Use With (Application) Used for Histological/Cytological Staining
Quantity 4 L