missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains
Description
Prepared with stain certified by the Biological Stain Commission
Specifications
Specifications
| Type | Harris Hematoxylin - Mercury-free, Non-acidified |
| Format | Liquid |
| For Use With (Application) | Used for Histological/Cytological Staining |
| Quantity | 4 L |