missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains

Catalog No. 23245654
Change view
Click to view available options
Quantity:
475 mL
500 mL
32 oz.
1 L
1 gal.
4 L
Type:
Alcoholic Eosin Y Stain
Bluing Reagent
Cyto-Quik Combo EA/OG Stain
Eosin Y Intensified
Gill Hematoxylin (1X)
Gill Hematoxylin (2X)
Gill Hematoxylin (3X)
Gomori Trichrome
Harris Hematoxylin
Harris Hematoxylin - Mercury-free
Harris Hematoxylin - Mercury-free, Non-acidified
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23245654 475 mL Gill Hematoxylin (2X)
23245651 4 L Harris Hematoxylin - Mercury-free
23245655 4 L Gill Hematoxylin (2X)
23245653 4 L Gill Hematoxylin (1X)
23245658 1 L Alcoholic Eosin Y Stain
23245657 4 L Gill Hematoxylin (3X)
23245681 4 L Bluing Reagent
23245678 4 L Harris Hematoxylin
23245677 475 mL Harris Hematoxylin - Mercury-free
23245679 500 mL Gomori Trichrome
23245697 4 L Harris Hematoxylin - Mercury-free, Non-acidified
23245827 4 L Alcoholic Eosin Y Stain
23310349 1 gal. Cyto-Quik Combo EA/OG Stain
23314631 1 gal. Eosin Y Intensified
23314630 32 oz. Eosin Y Intensified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 23245654 Supplier Fisher Healthcare™ Supplier No. 245654
Only null left

Manufactured with strict quality control to ensure lot-to-lot uniformity

Prepared with stain certified by the Biological Stain Commission

Specifications

Boiling Point 121°C
Color Maroon
Concentration 2X
Type Gill Hematoxylin (2X)
Format Liquid
For Use With (Application) Used for Histological/Cytological Staining
Quantity 475 mL