missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Healthcare™ PROTOCOL™ Hema-Quik™ and Hema-Quik II Stain Solutions
Description
Stains for manual and automated staining methods.
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Remain stable for a minimum of 12 months when stored at room temperature
- Allow varied staining times to suit individual preferences
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
Specifications
| Quantity | 1 gal. |
| Storage Requirements | Room Temperature |
| Boiling Point | 65°C |
| Format | Liquid |
| Type | Hema-Quik Stain Solution |
| For Use With (Application) | For Wright stain quality, fast single solution manual staining |
| Color | Dark Blue |