Learn More
Fisher Healthcare™ PROTOCOL™ Hema-G Stains for Geometric Data Hemastainer™
Quality-control tested on human blood smears to ensure proper staining and absence of precipitate
Supplier:  Fisher Healthcare™ 247056
 View more versions of this product
Catalog No. 23247056
Only null left
Description
Description
Stains for manual and automated staining methods
  • All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
  • Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
  • Carefully balanced for optimum staining pH
  • Remain stable for a minimum of 12 months when stored at room temperature
  • Allow varied staining times to suit individual preferences
  • Packaged in durable plastic bottles for safety and easy handling
  • Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
Chemical Identifiers
Room Temperature
Geometric data Hemastainer™
Specifications
1 gal.
65°C
Hema-G Wright Stain, Regular
Dark Purple
Room Temperature
Liquid
Geometric data Hemastainer™
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1 Year