missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Hema 3™ Fixative and Solutions

Catalog No. 23122952
Change view
Click to view available options
Product Type:
Hema 3 Fixative
Hema 3 Solution I
Hema 3 Solution II
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23122952 Hema 3 Solution II
23122929 Hema 3 Fixative
23122937 Hema 3 Solution I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23122952 Supplier Fisher Healthcare™ Supplier No. 122952
Edge

missing translation for 'validMainframeMsg'

For 30-second, three-step staining comparable to the Wright-Giemsa method

Specifications

Quantity 1 gal.
Boiling Point 100°C
Format Liquid
For Use With (Application) For manual staining
Color Blue
Product Type Hema 3 Solution II