Learn More
Fisher Healthcare™ Pinnacle Portfolio Bluing Reagent Stain
Catalog No. 22220106
Change view
Click to view available options
Quantity:
4 x 1 gal.
Catalog No. Quantity
22220106 4 x 1 gal.
1 options
Catalog No. 22220106 Supplier Fisher Healthcare™ Supplier No. 220106
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ensures crisp nuclear detail
  • Designed as a system
  • Each component has added value and benefits that reduces cost per slide
  • Unique chemical formulae of the Pinnacle Portfolio promote long-term chemical stability, which extends the usable life of each component
  • Subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity
Bluing Reagent Features:
  • Buffered to ensure the proper alkalinity (pH = 8.0)
  • Requires only a one-minute rinse in water versus conventional bluing agents which require up to five minutes of rinsing
  • Does not allow for tissue sections to be washed off the slide during the remainder of the staining process
  • Premixed and ready-to-use
Specifications
Quantity 4 x 1 gal.
Product Type Bluing Reagent