Learn More
Fisher Healthcare™ PROTOCOL™ Individual Gram Stains
Manufactured with traditional formulations used by most laboratories
Supplier:  Fisher Healthcare™ 291475
 View more versions of this product
Catalog No. 23-291475
Only null left
Explore available promotions
Description
Description
  • Decolorizer available in two different acetone/isopropanol ratios: 1:4 and 1:1
  • Dispenser caps are provided with the 8 oz. and 16 oz. sizes
  • Gallon-size Iodine is available as traditional diluent/concentrate or as ready-to-use Lugol's
A $10.00 service charge will be added when ordering Cat. No. 23-291-473 online.
Order Info
All sizes come in polyethylene bottles, except for Lugol's Iodine, which comes in a glass bottle.
Specifications
Specifications
Gram Decolorizer
Dispenser cap
Clear
Ethanol, Methanol, Acetone
8 oz.
Bottle
56.1° to 82°C
Gram Stain
Product Suggestions
Product Suggestions