missing translation for 'onlineSavingsMsg'
Learn More

Fisher Container™ Precision II™ High Clarity Cleanroom Tubing

Cleanroom tubing to meet your needs.

Supplier:  Fisher Container™ 1N400180

Encompass

 View more versions of this product

Catalog No. 19351165


Only null left

Description

Description

  • Bulk packed with Certificates of Conformance
  • Double bagged
  • Tubing optionally provided on plastic cores
  • Clear
  • Amine-free

Order Info

All tubing is 500 ft. long (152.4m).

Specifications

Chemical Identifiers

500 ft.
152.4 m

Specifications

18 in.
500 ft.
152.4 m
4 mil
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions