missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Yeast Extract (Granulated)

Catalog No. p-4245067
Change view
Click to view available options
Quantity:
500 g
2000 g
5 kg
Packaging Type:
Poly Bottle
Poly Pail
Media Type:
Yeast Extract
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type Media Type
BP9727500 500 g Poly Bottle Yeast Extract
BP97272 2000 g Poly Bottle Yeast Extract
BP97275 5 kg Poly Pail Yeast Extract
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP9727500 Supplier Fisher BioReagents™ Supplier No. BP9727 500
Only null left

Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins

  • Water-soluble portion of autolyzed yeast containing vitamin B complex
  • Used in preparing microbiological culture media
  • CAS: 8013-01-2

Specifications

Form Dehydrated
Media Type Yeast Extract
Product Type Microbiology Media
For Use With (Application) Used for preparing microbiological culture media
Includes Vitamin B complex
Packaging Type Poly Bottle
Quantity 500 g